siRNA Details
Gene Symbol: TMEM230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009923.1 | NP_001009923.1 | 3 | 327 | ||
NM_001009924.1 | NP_001009924.1 | 3 | 545 | |||
NM_001009925.1 | NP_001009925.1 | 2 | 221 | |||
NM_014145.4 | NP_054864.3 | 2 | 452 | |||
XM_005260713.3 | XP_005260770.1 | 2 | 419 | |||
XM_011529227.2 | XP_011527529.1 | 2 | 442 | |||
XM_011529229.2 | XP_011527531.1 | 3 | 467 | |||
XM_017027834.1 | XP_016883323.1 | 2 | 200 | |||
XM_017027835.1 | XP_016883324.1 | 2 | 221 | |||
XM_017027836.1 | XP_016883325.1 | 2 | 204 | |||
GenBank mRNA | AF161392.1 | - | 3 | 310 | ||
AK055700.1 | - | 3 | 501 | |||
AK309823.1 | - | 2 | 221 | |||
AK315606.1 | - | 2 | 211 | |||
AY359115.1 | - | 2 | 344 | |||
BC009768.1 | - | 2 | 205 | 4092724 | ||
BC009769.1 | - | 2 | 211 | 4132400 | ||
BC009770.1 | - | 2 | 205 | 4249231 | ||
BC011990.1 | - | 2 | 192 | 3857616 | ||
BC015113.1 | - | 2 | 156 | 4049350 | ||
BC070212.1 | - | 2 | 221 | 4687557 | ||
BC110408.1 | - | 2 | 183 | 5740520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
