siRNA Details
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 5, 6 | 598 | ||
NM_015874.4 | NP_056958.3 | 4, 5 | 545 | |||
NM_203283.2 | NP_976028.1 | 4, 5 | 482 | |||
NM_203284.2 | NP_976029.1 | 5, 6 | 728 | |||
XM_005248161.3 | XP_005248218.1 | 5, 6 | 612 | |||
XM_011513840.2 | XP_011512142.1 | 5, 6 | 1004 | |||
XM_017008170.1 | XP_016863659.1 | 4, 5 | 401 | |||
XM_017008171.1 | XP_016863660.1 | 4, 5 | 380 | |||
XM_017008172.1 | XP_016863661.1 | 5, 6 | 543 | |||
XM_017008173.1 | XP_016863662.1 | 5, 6 | 821 | |||
XM_017008174.1 | XP_016863663.1 | 5, 6 | 637 | |||
XM_017008175.1 | XP_016863664.1 | 5, 6 | 553 | |||
GenBank mRNA | AK302230.1 | - | 4, 5 | 335 | ||
AK303244.1 | - | 4, 5 | 394 | |||
AK311419.1 | - | 4, 5 | 381 | |||
BC020780.1 | - | 4, 5 | 482 | 4718839 | ||
BC064976.1 | - | 5, 6 | 503 | 6152924 | ||
BM472788.1 | - | 552 | ||||
BM555733.1 | - | 5, 6 | 706 | |||
BQ216033.1 | - | 4, 5 | 526 | |||
D14041.1 | - | 2, 3 | 375 | |||
DC361185.1 | - | 4, 5 | 546 | |||
L07872.1 | - | 4, 5 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
