siRNA Details
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 7, 8 | 913 | ||
NM_015874.4 | NP_056958.3 | 6, 7 | 860 | |||
NM_203283.2 | NP_976028.1 | 6, 7 | 797 | |||
NM_203284.2 | NP_976029.1 | 7, 8 | 1043 | |||
XM_005248161.3 | XP_005248218.1 | 7, 8 | 927 | |||
XM_011513840.2 | XP_011512142.1 | 7, 8 | 1319 | |||
XM_017008170.1 | XP_016863659.1 | 6, 7 | 716 | |||
XM_017008171.1 | XP_016863660.1 | 6, 7 | 695 | |||
XM_017008172.1 | XP_016863661.1 | 7, 8 | 858 | |||
XM_017008173.1 | XP_016863662.1 | 7, 8 | 1136 | |||
XM_017008174.1 | XP_016863663.1 | 7, 8 | 952 | |||
XM_017008175.1 | XP_016863664.1 | 7, 8 | 868 | |||
GenBank mRNA | AK126357.1 | - | 1, 2 | 3406 | ||
AK302230.1 | - | 6, 7 | 650 | |||
AK303244.1 | - | 6, 7 | 709 | |||
AK311419.1 | - | 6, 7 | 696 | |||
BC020780.1 | - | 6, 7 | 797 | 4718839 | ||
BC064976.1 | - | 7, 8 | 818 | 6152924 | ||
L07872.1 | - | 6, 7 | 755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
