siRNA Details
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 6 | 644 | ||
NM_015874.4 | NP_056958.3 | 5 | 591 | |||
NM_203283.2 | NP_976028.1 | 5 | 528 | |||
NM_203284.2 | NP_976029.1 | 6 | 774 | |||
XM_005248161.3 | XP_005248218.1 | 6 | 658 | |||
XM_011513840.2 | XP_011512142.1 | 6 | 1050 | |||
XM_017008170.1 | XP_016863659.1 | 5 | 447 | |||
XM_017008171.1 | XP_016863660.1 | 5 | 426 | |||
XM_017008172.1 | XP_016863661.1 | 6 | 589 | |||
XM_017008173.1 | XP_016863662.1 | 6 | 867 | |||
XM_017008174.1 | XP_016863663.1 | 6 | 683 | |||
XM_017008175.1 | XP_016863664.1 | 6 | 599 | |||
GenBank mRNA | AK302230.1 | - | 5 | 381 | ||
AK303244.1 | - | 5 | 440 | |||
AK311419.1 | - | 5 | 427 | |||
BC020780.1 | - | 5 | 528 | 4718839 | ||
BC064976.1 | - | 6 | 549 | 6152924 | ||
BE269143.1 | - | 6 | 554 | |||
BQ216033.1 | - | 5 | 572 | |||
L07872.1 | - | 5 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
