siRNA Details
Gene Symbol: ZSCAN32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284527.1 | NP_001271456.1 | 6 | 1248 | ||
NM_001284528.1 | NP_001271457.1 | 3 | 863 | |||
NM_001284529.1 | NP_001271458.1 | 2 | 310 | |||
NM_001324340.1 | NP_001311269.1 | 6 | 1298 | |||
NM_001324341.1 | NP_001311270.1 | 5 | 1082 | |||
NM_001324342.1 | NP_001311271.1 | 5 | 1203 | |||
NM_001324343.1 | NP_001311272.1 | 5 | 695 | |||
NM_001324344.1 | NP_001311273.1 | 5 | 1132 | |||
NM_001324345.1 | NP_001311274.1 | 4 | 1037 | |||
NM_001324346.1 | NP_001311275.1 | 4 | 1029 | |||
NM_017810.3 | NP_060280.2 | 5 | 1146 | |||
XM_005255403.2 | XP_005255460.1 | 4 | 987 | |||
XM_011522555.2 | XP_011520857.1 | 6 | 1152 | |||
XM_017023371.1 | XP_016878860.1 | 6 | 1102 | |||
XM_017023372.1 | XP_016878861.1 | 4 | 891 | |||
XM_017023373.1 | XP_016878862.1 | 4 | 840 | |||
GenBank mRNA | AK056463.1 | - | 3 | 792 | ||
AK291213.1 | - | 6 | 1194 | |||
AK299070.1 | - | 4 | 598 | |||
AK301586.1 | - | 2 | 239 | |||
AK303338.1 | - | 4 | 448 | |||
AY261676.1 | - | 5 | 1106 | |||
AY279347.1 | - | 5 | 1128 | |||
AY279348.1 | - | 5 | 1128 | |||
BC002859.2 | - | 6 | 1145 | 3638050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
