siRNA Details
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256737.1 | NP_001243666.1 | 1 | 148 | ||
NM_001256739.1 | NP_001243668.1 | 1 | 148 | |||
NM_001256740.1 | NP_001243669.1 | 1 | 148 | |||
NM_024034.4 | NP_076939.3 | 1 | 148 | |||
NR_046353.1 | 1 | 148 | ||||
GenBank mRNA | AF277566.1 | - | 1 | 103 | ||
AK056251.1 | - | 1 | 138 | |||
AK225166.1 | - | 1 | 138 | |||
AK293484.1 | - | 1 | 98 | |||
AK297348.1 | - | 1 | 50 | |||
AK299259.1 | - | 1 | 98 | |||
AK299700.1 | - | 1 | 97 | |||
BC000199.1 | - | 1 | 124 | 3352559 | ||
BC009014.1 | - | 1 | 87 | 4155653 | ||
BP307776.1 | - | 1 | 148 | |||
BU165271.1 | - | 1 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
