siRNA Details
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256737.1 | NP_001243666.1 | 2 | 279 | ||
NM_001256738.1 | NP_001243667.1 | 2 | 126 | |||
NM_001256739.1 | NP_001243668.1 | 2 | 279 | |||
NM_001256740.1 | NP_001243669.1 | 2 | 279 | |||
NM_024034.4 | NP_076939.3 | 2 | 279 | |||
GenBank mRNA | AF123764.1 | - | 2 | 175 | ||
AF123765.1 | - | 2 | 218 | |||
AF277566.1 | - | 2 | 234 | |||
AK056251.1 | - | 2 | 269 | |||
AK225166.1 | - | 2 | 269 | |||
AK293484.1 | - | 2 | 229 | |||
AK299259.1 | - | 3 | 191 | |||
AK299700.1 | - | 2 | 228 | |||
BC000199.1 | - | 2 | 255 | 3352559 | ||
BC009014.1 | - | 2 | 218 | 4155653 | ||
BP307776.1 | - | 2 | 279 | |||
BU165271.1 | - | 2 | 254 | |||
DA528340.1 | - | 2 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
