siRNA Details
Gene Symbol: TMEM200B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003682.3 | NP_001003682.1 | 2 | 272 | ||
NM_001171868.1 | NP_001165339.1 | 2 | 356 | |||
XM_011541457.2 | XP_011539759.1 | 2 | 397 | |||
GenBank mRNA | AL110282.1 | - | 1 | 563 | ||
BC064375.1 | - | 2 | 258 | 5722634 | ||
BC064538.1 | - | 1 | 197 | |||
BC082989.1 | - | 2 | 271 | 5763085 | ||
BC110336.1 | - | 2 | 356 | 5753003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
