siRNA Details
Gene Symbol: H2AFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002105.2 | NP_002096.1 | 1 | 191 | ||
GenBank mRNA | BC004915.2 | - | 1 | 161 | 3537648 | |
BC011694.2 | - | 1 | 171 | 3139343 | ||
BC013416.2 | - | 1 | 156 | 3534359 | ||
CR457079.1 | - | 1 | 118 | |||
X14850.1 | - | 1 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
