siRNA Details
Gene Symbol: ELK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114123.2 | NP_001107595.1 | 3 | 420 | ||
NM_001257168.1 | NP_001244097.1 | 3 | 420 | |||
NM_005229.4 | NP_005220.2 | 2 | 314 | |||
XM_017029339.1 | XP_016884828.1 | 1 | 135 | |||
GenBank mRNA | AB016193.1 | - | 2 | 198 | ||
AB209047.1 | - | 2 | 287 | |||
AF000672.1 | - | 3 | 413 | |||
AK312984.1 | - | 3 | 275 | |||
BC048296.1 | - | 2 | 176 | |||
BC056150.1 | - | 1 | 144 | 6503270 | ||
CR542252.1 | - | 1 | 98 | |||
M25269.1 | - | 3 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
