siRNA Details
Gene Symbol: TNNI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000363.4 | NP_000354.4 | 8 | 825 | ||
GenBank mRNA | AK223451.1 | - | 8 | 825 | ||
EU747720.1 | - | 8 | 825 | |||
M64247.1 | - | 8 | 690 | |||
X54163.1 | - | 8 | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
