siRNA Details
Gene Symbol: TNNI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000363.4 | NP_000354.4 | 7, 8 | 691 | ||
GenBank mRNA | AK223451.1 | - | 7, 8 | 691 | ||
BC096165.1 | - | 7, 8 | 617 | 40003354 | ||
BC096166.1 | - | 7, 8 | 617 | 40003355 | ||
BC096167.3 | - | 7, 8 | 617 | 40003357 | ||
BC099631.3 | - | 7, 8 | 617 | 40003359 | ||
BT019517.1 | - | 5, 6 | 548 | |||
CR541985.1 | - | 5, 6 | 548 | |||
CR542012.1 | - | 5, 6 | 548 | |||
EU747720.1 | - | 7, 8 | 691 | |||
M64247.1 | - | 7, 8 | 556 | |||
X54163.1 | - | 7, 8 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
