siRNA Details
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077427.3 | NP_001070895.1 | 2 | 485 | ||
NM_001321234.1 | NP_001308163.1 | 3 | 447 | |||
NM_001321235.1 | NP_001308164.1 | 2 | 608 | |||
NM_144586.6 | NP_653187.3 | 2 | 574 | |||
XM_011510546.2 | XP_011508848.1 | 2 | 1127 | |||
GenBank mRNA | AK075487.1 | - | 2 | 409 | ||
AK094501.1 | - | 3 | 350 | |||
AK123029.1 | - | 3 | 307 | |||
AK130005.1 | - | 3 | 286 | |||
AK309668.1 | - | 2 | 485 | |||
AK309849.1 | - | 3 | 294 | |||
AY358628.1 | - | 2 | 256 | |||
BC017318.2 | - | 2 | 387 | 3641660 | ||
BE386060.1 | - | 2 | 331 | |||
BP276862.1 | - | 2 | 438 | |||
CB110132.1 | - | 2 | 403 | |||
DA776269.1 | - | 2 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
