siRNA Details
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077427.3 | NP_001070895.1 | 2, 3 | 533 | ||
NM_001321234.1 | NP_001308163.1 | 3, 4 | 495 | |||
NM_001321235.1 | NP_001308164.1 | 2, 3 | 656 | |||
NM_144586.6 | NP_653187.3 | 2, 3 | 622 | |||
GenBank mRNA | AK075487.1 | - | 2, 3 | 457 | ||
AK094501.1 | - | 3, 4 | 398 | |||
AK123029.1 | - | 3, 4 | 355 | |||
AK130005.1 | - | 3, 4 | 334 | |||
AK309668.1 | - | 2, 3 | 533 | |||
AK309849.1 | - | 3, 4 | 342 | |||
AY358628.1 | - | 2, 3 | 304 | |||
BC017318.2 | - | 2, 3 | 435 | 3641660 | ||
BE386060.1 | - | 2, 3 | 379 | |||
BP276862.1 | - | 2, 3 | 486 | |||
DA776269.1 | - | 2, 3 | 524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
