siRNA Details
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 17 | 1732 | ||
NM_018193.2 | NP_060663.2 | 17 | 1732 | |||
XM_011521756.2 | XP_011520058.1 | 17 | 1756 | |||
XM_011521757.2 | XP_011520059.1 | 17 | 1752 | |||
XM_011521764.2 | XP_011520066.1 | 17 | 1756 | |||
XM_011521767.2 | XP_011520069.1 | 17 | 1726 | |||
GenBank mRNA | AB058697.1 | - | 3 | 1656 | ||
AK001581.1 | - | 11 | 1121 | |||
AK027564.1 | - | 6 | 619 | |||
AK055176.1 | - | 17 | 1691 | |||
BC140769.1 | - | 17 | 1689 | 9021637 | ||
BC144483.1 | - | 17 | 1689 | 9053010 | ||
EF469766.1 | - | 16 | 1642 | |||
EF567077.1 | - | 16 | 1642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
