siRNA Details
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 26, 27 | 2981 | ||
NM_018193.2 | NP_060663.2 | 25, 26 | 2801 | |||
XM_011521756.2 | XP_011520058.1 | 26, 27 | 3005 | |||
XM_011521757.2 | XP_011520059.1 | 26, 27 | 3001 | |||
XM_011521764.2 | XP_011520066.1 | 25, 26 | 2825 | |||
XM_011521767.2 | XP_011520069.1 | 26, 27 | 2975 | |||
GenBank mRNA | AB058697.1 | - | 12, 13 | 2905 | ||
AK001581.1 | - | 19, 20 | 2190 | |||
AK027564.1 | - | 14, 15 | 1688 | |||
AK055176.1 | - | 25, 26 | 2760 | |||
BC021859.1 | - | 3, 4 | 323 | |||
BC140769.1 | - | 26, 27 | 2938 | 9021637 | ||
BC144483.1 | - | 26, 27 | 2938 | 9053010 | ||
EF469766.1 | - | 25, 26 | 2891 | |||
EF567077.1 | - | 25, 26 | 2891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
