siRNA Details
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 27, 28 | 3104 | ||
NM_018193.2 | NP_060663.2 | 26, 27 | 2924 | |||
XM_011521756.2 | XP_011520058.1 | 27, 28 | 3128 | |||
XM_011521757.2 | XP_011520059.1 | 27, 28 | 3124 | |||
XM_011521764.2 | XP_011520066.1 | 26, 27 | 2948 | |||
XM_011521767.2 | XP_011520069.1 | 27, 28 | 3098 | |||
GenBank mRNA | AB058697.1 | - | 13, 14 | 3028 | ||
AK001581.1 | - | 20, 21 | 2313 | |||
AK027564.1 | - | 15, 16 | 1811 | |||
AK055176.1 | - | 26, 27 | 2883 | |||
BC021859.1 | - | 4, 5 | 446 | |||
BC140769.1 | - | 27, 28 | 3061 | 9021637 | ||
BC144483.1 | - | 27, 28 | 3061 | 9053010 | ||
EF469766.1 | - | 26, 27 | 3014 | |||
EF567077.1 | - | 26, 27 | 3014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
