siRNA Details
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270764.1 | NP_001257693.1 | 2 | 803 | ||
NM_001270765.1 | NP_001257694.1 | 2 | 803 | |||
NM_014863.3 | NP_055678.2 | 2 | 765 | |||
NM_015892.4 | NP_056976.2 | 2 | 765 | |||
XM_005269891.3 | XP_005269948.1 | 2 | 881 | |||
XM_005269892.4 | XP_005269949.1 | 2 | 880 | |||
XM_005269893.3 | XP_005269950.1 | 2 | 880 | |||
XM_005269894.4 | XP_005269951.1 | 2 | 880 | |||
XM_006717891.3 | XP_006717954.1 | 2 | 1198 | |||
XM_011539857.1 | XP_011538159.1 | 2 | 880 | |||
XM_017016319.1 | XP_016871808.1 | 2 | 717 | |||
XM_017016320.1 | XP_016871809.1 | 2 | 755 | |||
XM_017016321.1 | XP_016871810.1 | 2 | 801 | |||
GenBank mRNA | AB011170.1 | - | 2 | 733 | ||
AB062423.1 | - | 1 | 152 | |||
AF026477.1 | - | 1 | 358 | |||
AK294961.1 | - | 2 | 755 | |||
AK299546.1 | - | 2 | 794 | |||
BC027908.1 | - | 2 | 708 | 5204516 | ||
BC050540.1 | - | 2 | 783 | 5741278 | ||
BC075813.1 | - | 1 | 459 | 5213653 | ||
CR749804.1 | - | 2 | 886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
