siRNA Details
Gene Symbol: MEF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271629.1 | NP_001258558.1 | 9 | 1419 | ||
NM_005920.3 | NP_005911.1 | 10 | 1721 | |||
XM_005245169.4 | XP_005245226.1 | 10 | 1703 | |||
XM_005245170.3 | XP_005245227.1 | 10 | 1728 | |||
XM_006711330.3 | XP_006711393.1 | 10 | 1432 | |||
XM_006711332.3 | XP_006711395.1 | 10 | 1716 | |||
XM_006711333.2 | XP_006711396.1 | 9 | 1695 | |||
XM_006711334.3 | XP_006711397.1 | 9 | 1693 | |||
XM_011509569.2 | XP_011507871.1 | 10 | 1753 | |||
XM_017001314.1 | XP_016856803.1 | 9 | 1837 | |||
XM_017001315.1 | XP_016856804.1 | 9 | 1432 | |||
GenBank mRNA | AB210005.1 | - | 10 | 1635 | ||
AK308641.1 | - | 10 | 1621 | |||
BC032479.1 | - | 7 | 1270 | |||
BC040949.1 | - | 9 | 1419 | 6044434 | ||
BC054520.1 | - | 10 | 1616 | 6142751 | ||
L16794.1 | - | 10 | 1421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
