siRNA Details
Gene Symbol: MEF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271629.1 | NP_001258558.1 | 2 | 249 | ||
NM_005920.3 | NP_005911.1 | 2 | 530 | |||
XM_005245169.4 | XP_005245226.1 | 2 | 512 | |||
XM_005245170.3 | XP_005245227.1 | 2 | 537 | |||
XM_006711330.3 | XP_006711393.1 | 2 | 241 | |||
XM_006711332.3 | XP_006711395.1 | 2 | 528 | |||
XM_006711333.2 | XP_006711396.1 | 2 | 525 | |||
XM_006711334.3 | XP_006711397.1 | 2 | 526 | |||
XM_011509569.2 | XP_011507871.1 | 2 | 562 | |||
XM_017001314.1 | XP_016856803.1 | 2 | 667 | |||
XM_017001315.1 | XP_016856804.1 | 2 | 262 | |||
GenBank mRNA | AB210005.1 | - | 2 | 444 | ||
AK308641.1 | - | 2 | 430 | |||
BC032479.1 | - | 2 | 441 | |||
BC040949.1 | - | 2 | 249 | 6044434 | ||
BC054520.1 | - | 2 | 425 | 6142751 | ||
L16794.1 | - | 2 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
