siRNA Details
Gene Symbol: MINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024937.3 | NP_001020108.1 | 27 | 3470 | ||
NM_001321236.1 | NP_001308165.1 | 27 | 3535 | |||
NM_015716.4 | NP_056531.1 | 26 | 3419 | |||
NM_153827.4 | NP_722549.2 | 27 | 3530 | |||
NM_170663.4 | NP_733763.1 | 27 | 3443 | |||
XM_006721531.2 | XP_006721594.1 | 28 | 3617 | |||
XM_006721532.2 | XP_006721595.1 | 28 | 3614 | |||
XM_006721536.2 | XP_006721599.1 | 27 | 3506 | |||
XM_011523906.1 | XP_011522208.1 | 27 | 3521 | |||
XM_017024704.1 | XP_016880193.1 | 28 | 3608 | |||
XM_017024705.1 | XP_016880194.1 | 28 | 3557 | |||
XM_017024706.1 | XP_016880195.1 | 27 | 3503 | |||
XM_017024707.1 | XP_016880196.1 | 28 | 3418 | |||
XM_017024708.1 | XP_016880197.1 | 26 | 3268 | |||
GenBank mRNA | AB035698.1 | - | 26 | 3175 | ||
AB041926.1 | - | 27 | 3383 | |||
AB209450.1 | - | 26 | 4068 | |||
AF218033.1 | - | 5 | 548 | |||
AK096610.1 | - | 2 | 1104 | |||
AL157418.1 | - | 22 | 3082 | |||
AY775058.1 | - | 27 | 3226 | |||
BC034673.1 | - | 27 | 3482 | 4384442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
