siRNA Details
Gene Symbol: MINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024937.3 | NP_001020108.1 | 27, 28 | 3546 | ||
NM_001321236.1 | NP_001308165.1 | 27, 28 | 3611 | |||
NM_015716.4 | NP_056531.1 | 26, 27 | 3495 | |||
NM_153827.4 | NP_722549.2 | 27, 28 | 3606 | |||
NM_170663.4 | NP_733763.1 | 27, 28 | 3519 | |||
XM_006721531.2 | XP_006721594.1 | 28, 29 | 3693 | |||
XM_006721532.2 | XP_006721595.1 | 28, 29 | 3690 | |||
XM_006721536.2 | XP_006721599.1 | 27, 28 | 3582 | |||
XM_011523906.1 | XP_011522208.1 | 27, 28 | 3597 | |||
XM_017024704.1 | XP_016880193.1 | 28, 29 | 3684 | |||
XM_017024705.1 | XP_016880194.1 | 28, 29 | 3633 | |||
XM_017024706.1 | XP_016880195.1 | 27, 28 | 3579 | |||
XM_017024707.1 | XP_016880196.1 | 28, 29 | 3494 | |||
XM_017024708.1 | XP_016880197.1 | 26, 27 | 3344 | |||
GenBank mRNA | AB035698.1 | - | 26, 27 | 3251 | ||
AB041926.1 | - | 27, 28 | 3459 | |||
AB209450.1 | - | 26, 27 | 4144 | |||
AF218033.1 | - | 5, 6 | 624 | |||
AL157418.1 | - | 22, 23 | 3158 | |||
AY775058.1 | - | 27, 28 | 3302 | |||
BC034673.1 | - | 27, 28 | 3558 | 4384442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
