siRNA Details
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142314.1 | NP_001135786.1 | 2 | 632 | ||
NM_001321519.1 | NP_001308448.1 | 2 | 632 | |||
NM_080876.3 | NP_543152.1 | 2 | 632 | |||
GenBank mRNA | AB038770.1 | - | 2 | 313 | ||
AB063186.1 | - | 2 | 637 | |||
AB063187.1 | - | 2 | 637 | |||
AF486808.1 | - | 2 | 401 | |||
AK314078.1 | - | 2 | 442 | |||
BC035000.1 | - | 2 | 439 | 5269106 | ||
BC093958.1 | - | 2 | 355 | 7939803 | ||
BC112005.1 | - | 2 | 367 | 8327473 | ||
BX647818.1 | - | 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
