siRNA Details
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300918.1 | NP_001287847.1 | 2 | 993 | ||
NM_001300919.1 | NP_001287848.1 | 2 | 993 | |||
NM_003483.4 | NP_003474.1 | 2 | 993 | |||
NM_003484.1 | NP_003475.1 | 2 | 993 | |||
XM_017019989.1 | XP_016875478.1 | 2 | 642 | |||
GenBank mRNA | AB209853.1 | - | 2 | 555 | ||
AF533652.1 | - | 1 | 91 | |||
AF533653.1 | - | 85 | ||||
AF533654.1 | - | 1 | 85 | |||
AI290477.1 | - | 1 | 71 | |||
AK311399.1 | - | 2 | 642 | |||
AY601861.1 | - | 2 | 993 | |||
AY601862.1 | - | 2 | 993 | |||
AY601863.1 | - | 2 | 993 | |||
AY601864.1 | - | 2 | 993 | |||
AY601865.1 | - | 2 | 993 | |||
AY601866.1 | - | 2 | 993 | |||
AY601867.1 | - | 2 | 993 | |||
U28749.1 | - | 2 | 993 | |||
X92518.1 | - | 2 | 1492 | |||
Z31595.1 | - | 2 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
