siRNA Details
Gene Symbol: LINC00521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024182.1 | 2 | 198 | |||
NR_024183.1 | 2 | 450 | ||||
NR_024184.1 | 2 | 450 | ||||
GenBank mRNA | AK097741.1 | - | 2 | 445 | ||
BC021728.2 | - | 2 | 449 | |||
BC031252.1 | - | 2 | 198 | |||
BI458986.1 | - | 2 | 454 | |||
DB446670.1 | - | 2 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
