siRNA Details
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 12 | 2605 | ||
GenBank mRNA | AK091918.1 | - | 12 | 2606 | ||
AK094502.1 | - | 2 | 563 | |||
BC101211.1 | - | 2 | 563 | |||
BC101212.1 | - | 2 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
