siRNA Details
Gene Symbol: MOV10L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164104.1 | NP_001157576.1 | 23 | 3252 | ||
NM_001164105.1 | NP_001157577.1 | 23 | 3329 | |||
NM_001164106.1 | NP_001157578.1 | 5 | 637 | |||
NM_018995.2 | NP_061868.1 | 23 | 3252 | |||
XM_005261923.3 | XP_005261980.1 | 23 | 3530 | |||
XM_011530696.1 | XP_011528998.1 | 23 | 3252 | |||
XM_011530697.1 | XP_011528999.1 | 23 | 3530 | |||
XM_011530698.1 | XP_011529000.1 | 22 | 3111 | |||
XM_011530699.1 | XP_011529001.1 | 23 | 3214 | |||
XM_011530700.2 | XP_011529002.1 | 23 | 3597 | |||
XM_011530701.1 | XP_011529003.1 | 23 | 3252 | |||
XM_011530702.1 | XP_011529004.1 | 22 | 3025 | |||
XM_011530703.1 | XP_011529005.1 | 16 | 2100 | |||
XM_017028833.1 | XP_016884322.1 | 23 | 3252 | |||
XM_017028835.1 | XP_016884324.1 | 14 | 1666 | |||
XM_017028836.1 | XP_016884325.1 | 14 | 1623 | |||
GenBank mRNA | AF285604.1 | - | 23 | 3196 | ||
AK000033.1 | - | 5 | 610 | |||
AK090740.1 | - | 3 | 603 | |||
AK092675.1 | - | 17 | 2088 | |||
AK302401.1 | - | 23 | 3329 | |||
AL133068.2 | - | 13 | 1571 | |||
BC140944.1 | - | 24 | 3253 | 9056756 | ||
BC150137.1 | - | 23 | 3194 | 40015813 | ||
BC152539.1 | - | 23 | 3194 | 40009193 | ||
CR456466.1 | - | 23 | 3175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
