siRNA Details
Gene Symbol: MOV10L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164104.1 | NP_001157576.1 | 22 | 2992 | ||
NM_001164105.1 | NP_001157577.1 | 22 | 3069 | |||
NM_001164106.1 | NP_001157578.1 | 4 | 377 | |||
NM_018995.2 | NP_061868.1 | 22 | 2992 | |||
XM_005261923.3 | XP_005261980.1 | 22 | 3270 | |||
XM_011530696.1 | XP_011528998.1 | 22 | 2992 | |||
XM_011530697.1 | XP_011528999.1 | 22 | 3270 | |||
XM_011530698.1 | XP_011529000.1 | 21 | 2851 | |||
XM_011530699.1 | XP_011529001.1 | 22 | 2954 | |||
XM_011530700.2 | XP_011529002.1 | 22 | 3337 | |||
XM_011530701.1 | XP_011529003.1 | 22 | 2992 | |||
XM_011530702.1 | XP_011529004.1 | 21 | 2765 | |||
XM_011530703.1 | XP_011529005.1 | 15 | 1840 | |||
XM_017028833.1 | XP_016884322.1 | 22 | 2992 | |||
XM_017028835.1 | XP_016884324.1 | 13 | 1406 | |||
XM_017028836.1 | XP_016884325.1 | 13 | 1363 | |||
GenBank mRNA | AF285604.1 | - | 22 | 2936 | ||
AK000033.1 | - | 4 | 350 | |||
AK090740.1 | - | 2 | 343 | |||
AK092675.1 | - | 16 | 1828 | |||
AK302401.1 | - | 22 | 3069 | |||
AL133068.2 | - | 12 | 1311 | |||
BC140944.1 | - | 23 | 2993 | 9056756 | ||
BC150137.1 | - | 22 | 2934 | 40015813 | ||
BC152539.1 | - | 22 | 2934 | 40009193 | ||
BP382225.1 | - | 4 | 377 | |||
CR456466.1 | - | 22 | 2915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
