siRNA Details
Gene Symbol: LOC105369739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135028.1 | 3 | 330 | |||
GenBank mRNA | BX096171.1 | - | 3 | 330 |
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130914.1 | 3 | 354 | |||
GenBank mRNA | AK002104.1 | - | 2 | 1048 | ||
AK002154.1 | - | 2 | 275 | |||
AK023417.1 | - | 3 | 355 | |||
BC117434.1 | - | 2 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
