siRNA Details
Gene Symbol: DNAJC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307980.1 | NP_001294909.1 | 2 | 583 | ||
NM_001307981.1 | NP_001294910.1 | 2 | 583 | |||
NM_005528.3 | NP_005519.2 | 3 | 622 | |||
GenBank mRNA | AF012106.1 | - | 1 | 332 | ||
AF036874.1 | - | 2 | 229 | |||
BC010145.2 | - | 2 | 448 | |||
BC032137.2 | - | 2 | 257 | |||
BC044584.1 | - | 3 | 625 | 4811478 | ||
BC051008.1 | - | 2 | 105 | |||
BC053947.1 | - | 3 | 625 | |||
BC057849.1 | - | 2 | 455 | 30349314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
