siRNA Details
Gene Symbol: ZNF492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020855.2 | NP_065906.1 | 4 | 2646 | ||
GenBank mRNA | AB040906.1 | - | 4 | 2534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
