siRNA Details
Gene Symbol: KRTAP13-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181621.3 | NP_853652.1 | 1 | 460 | ||
GenBank mRNA | AB096938.1 | - | 1 | 450 | ||
BC093934.1 | - | 1 | 459 | 7939779 | ||
BC093936.1 | - | 1 | 460 | 7939781 | ||
BX104478.1 | - | 1 | 477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
