siRNA Details
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 6 | 1069 | ||
NM_052917.3 | NP_443149.2 | 8 | 1547 | |||
XM_011510537.2 | XP_011508839.1 | 8 | 1542 | |||
XM_011510538.2 | XP_011508840.1 | 6 | 954 | |||
XM_017003258.1 | XP_016858747.1 | 8 | 1396 | |||
XM_017003259.1 | XP_016858748.1 | 7 | 1121 | |||
XM_017003260.1 | XP_016858749.1 | 8 | 1542 | |||
XM_017003261.1 | XP_016858750.1 | 8 | 1406 | |||
XM_017003262.1 | XP_016858751.1 | 6 | 812 | |||
XM_017003263.1 | XP_016858752.1 | 6 | 954 | |||
GenBank mRNA | AB067505.1 | - | 6 | 847 | ||
AB078142.1 | - | 6 | 965 | |||
AJ505991.2 | - | 6 | 965 | |||
AK123152.1 | - | 1230 | ||||
AK131195.1 | - | 8 | 1492 | |||
AK308675.1 | - | 6 | 954 | |||
BC101031.1 | - | 6 | 965 | 40007710 | ||
BC101032.2 | - | 6 | 965 | 40007711 | ||
BC101033.1 | - | 6 | 965 | 40007712 | ||
BC101034.2 | - | 6 | 965 | 40007713 | ||
KJ534843.1 | - | 6 | 965 | |||
KJ535058.1 | - | 5 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
