siRNA Details
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 12 | 2096 | ||
NM_052917.3 | NP_443149.2 | 13 | 2406 | |||
XM_011510537.2 | XP_011508839.1 | 14 | 2599 | |||
XM_011510538.2 | XP_011508840.1 | 12 | 2011 | |||
XM_017003258.1 | XP_016858747.1 | 14 | 2453 | |||
XM_017003259.1 | XP_016858748.1 | 13 | 2178 | |||
XM_017003260.1 | XP_016858749.1 | 14 | 2569 | |||
XM_017003261.1 | XP_016858750.1 | 13 | 2265 | |||
XM_017003262.1 | XP_016858751.1 | 12 | 1839 | |||
XM_017003263.1 | XP_016858752.1 | 11 | 1813 | |||
GenBank mRNA | AB067505.1 | - | 11 | 1706 | ||
AK123152.1 | - | 2027 | ||||
AK131195.1 | - | 13 | 2351 | |||
AK308615.1 | - | 2 | 582 | |||
BC101031.1 | - | 11 | 1824 | 40007710 | ||
BC101032.2 | - | 12 | 1992 | 40007711 | ||
BC101033.1 | - | 11 | 1824 | 40007712 | ||
BC101034.2 | - | 11 | 1824 | 40007713 | ||
BG221433.1 | - | 2 | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
