siRNA Details
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 2 | 423 | ||
GenBank mRNA | AF312765.1 | - | 2 | 99 | ||
AJ457795.1 | - | 2 | 125 | |||
AY920734.1 | - | 2 | 89 | |||
AY920740.1 | - | 2 | 89 | |||
BC005382.1 | - | 2 | 182 | 3935644 | ||
BC062672.1 | - | 2 | 363 | 4047937 | ||
BC103961.1 | - | 2 | 122 | 40013988 | ||
BC121070.2 | - | 2 | 132 | 40119569 | ||
BC134418.1 | - | 2 | 132 | 40119568 | ||
BM554791.1 | - | 2 | 423 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
