siRNA Details
Gene Symbol: SLC22A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033396.1 | 4 | 711 | |||
XM_017017775.1 | XP_016873264.1 | 1 | 518 | |||
XM_017017776.1 | XP_016873265.1 | 1 | 518 | |||
GenBank mRNA | DY654625.1 | - | 4 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
