siRNA Details
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025109.1 | NP_001020280.1 | 5 | 947 | ||
NM_001773.2 | NP_001764.1 | 5 | 947 | |||
GenBank mRNA | AF202879.1 | - | 3 | 192 | ||
AF523361.1 | - | 5 | 760 | |||
AF538601.1 | - | 4 | 598 | |||
AK091890.1 | - | 2 | 664 | |||
AK291529.1 | - | 5 | 700 | |||
AK293030.1 | - | 5 | 724 | |||
AK294381.1 | - | 4 | 471 | |||
AK300835.1 | - | 6 | 764 | |||
BC039146.1 | - | 5 | 697 | 4746591 | ||
BX538076.1 | - | 2 | 875 | |||
M81104.1 | - | 5 | 946 | |||
S53910.1 | - | 5 | 779 | |||
S53911.1 | - | 5 | 779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
