siRNA Details
Gene Symbol: GPSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145638.2 | NP_001139110.2 | 8 | 1229 | ||
NM_015597.5 | NP_056412.5 | 8 | 1229 | |||
XM_011518498.2 | XP_011516800.1 | 8 | 1120 | |||
XM_017014598.1 | XP_016870087.1 | 7 | 941 | |||
GenBank mRNA | AL117478.1 | - | 2 | 69 | ||
AY173053.1 | - | 7 | 940 | |||
BC009979.1 | - | 8 | 1238 | 4121647 | ||
BC017353.1 | - | 8 | 1238 | 5021010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
