siRNA Details
Gene Symbol: APOBEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270411.1 | NP_001257340.1 | 8 | 1429 | ||
NM_004900.4 | NP_004891.4 | 8 | 1504 | |||
GenBank mRNA | AK024854.1 | - | 7 | 1785 | ||
BC053859.1 | - | 7 | 1376 | 6174285 | ||
BX114955.1 | - | 3 | 542 | |||
U61083.1 | - | 5 | 952 | |||
U61084.1 | - | 8 | 1493 |
Gene Symbol: APOBEC3A_B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193289.1 | NP_001180218.1 | 5 | 1070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
