siRNA Details
Gene Symbol: APOBEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270411.1 | NP_001257340.1 | 6, 7 | 995 | ||
NM_004900.4 | NP_004891.4 | 6, 7 | 1070 | |||
GenBank mRNA | AK024854.1 | - | 6, 7 | 1069 | ||
AY743217.1 | - | 6, 7 | 1015 | |||
BC053859.1 | - | 5, 6 | 942 | 6174285 | ||
BX114955.1 | - | 1, 2 | 108 | |||
CT841510.1 | - | 6, 7 | 965 | |||
KF651167.1 | - | 6, 7 | 1015 | |||
U61083.1 | - | 3, 4 | 518 | |||
U61084.1 | - | 6, 7 | 1059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
