siRNA Details
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004354.2 | NP_001004354.1 | 1 | 370 | ||
GenBank mRNA | BC053618.1 | - | 1 | 370 | 6143133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
