siRNA Details
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004354.2 | NP_001004354.1 | 1 | 459 | ||
GenBank mRNA | BC053618.1 | - | 1 | 459 | 6143133 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025980, AY046077, BC020293, BC048088, BC069891, and others.
Gene Ontology Categories:
