siRNA Details
Gene Symbol: ROMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080748.2 | NP_542786.1 | 3 | 379 | ||
XM_017027678.1 | XP_016883167.1 | 3 | 374 | |||
GenBank mRNA | AK000548.1 | - | 3 | 284 | ||
AM397244.1 | - | 3 | 278 | |||
AM397245.1 | - | 3 | 284 | |||
AM397246.1 | - | 2 | 359 | |||
BC008488.1 | - | 395 | 4133274 | |||
BC105281.1 | - | 3 | 288 | 6739248 | ||
GU727625.1 | - | 3 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
