siRNA Details
Gene Symbol: ROMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080748.2 | NP_542786.1 | 2, 3 | 271 | ||
XM_017027678.1 | XP_016883167.1 | 2, 3 | 266 | |||
GenBank mRNA | AK000548.1 | - | 2, 3 | 176 | ||
AM397244.1 | - | 2, 3 | 170 | |||
AM397245.1 | - | 2, 3 | 176 | |||
AM397246.1 | - | 1, 2 | 251 | |||
BC008488.1 | - | 287 | 4133274 | |||
BC105281.1 | - | 2, 3 | 180 | 6739248 | ||
GU727625.1 | - | 2, 3 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
