siRNA Details
Gene Symbol: ROMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080748.2 | NP_542786.1 | 3 | 431 | ||
XM_017027678.1 | XP_016883167.1 | 3 | 426 | |||
GenBank mRNA | AK000548.1 | - | 3 | 336 | ||
BC008488.1 | - | 447 | 4133274 | |||
BC105281.1 | - | 3 | 340 | 6739248 | ||
GU727625.1 | - | 3 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
