siRNA Details
Gene Symbol: IL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145659.3 | NP_663634.2 | 5 | 802 | ||
XM_011545780.2 | XP_011544082.1 | 5 | 1156 | |||
GenBank mRNA | BC062422.1 | - | 5 | 805 | 4703457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
