siRNA Details
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130807.2 | NP_570719.1 | 3 | 382 | ||
XM_011527683.2 | XP_011525985.1 | 3 | 459 | |||
GenBank mRNA | AJ580635.1 | - | 1 | 223 | ||
AK095471.1 | - | 3 | 382 | |||
AL832474.1 | - | 3 | 418 | |||
CR627409.1 | - | 3 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
