siRNA Details
Gene Symbol: PLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002667.4 | NP_002658.1 | 2 | 241 | ||
GenBank mRNA | AK311799.1 | - | 2 | 176 | ||
BC005269.1 | - | 2 | 176 | 3934199 | ||
BP257094.1 | - | 2 | 241 | |||
BP259965.1 | - | 2 | 225 | |||
M60411.1 | - | 1 | 88 | |||
M63603.1 | - | 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
