siRNA Details
Gene Symbol: ZBED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052923.1 | NP_443155.1 | 3 | 1708 | ||
XM_011514283.2 | XP_011512585.1 | 5 | 1148 | |||
XM_011514284.2 | XP_011512586.1 | 5 | 1104 | |||
XM_011514285.2 | XP_011512587.1 | 5 | 1070 | |||
XM_011514286.2 | XP_011512588.1 | 5 | 1081 | |||
XM_011514287.2 | XP_011512589.1 | 3 | 817 | |||
XM_011514288.2 | XP_011512590.1 | 2 | 605 | |||
XM_017010231.1 | XP_016865720.1 | 3 | 1141 | |||
GenBank mRNA | AB067512.1 | - | 2 | 558 | ||
AK056369.1 | - | 3 | 1663 | |||
AL832486.1 | - | 3 | 823 | |||
AY517631.1 | - | 3 | 1708 | |||
BC110834.1 | - | 3 | 1436 | 6499684 | ||
BC111741.1 | - | 3 | 1228 | 40038622 | ||
BC111742.1 | - | 3 | 1228 | 40038617 | ||
BX647791.1 | - | 3 | 1676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
