siRNA Details
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002125.3 | NP_002116.2 | 6 | 1054 | ||
XM_011514562.2 | XP_011512864.1 | 6 | 1227 | |||
GenBank mRNA | BC009234.2 | - | 6 | 1092 | 4054398 | |
M20429.1 | - | 6 | 1055 | |||
M32578.1 | - | 6 | 1110 | |||
M57648.1 | - | 6 | 1106 |
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093782.1 | - | 2250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
